missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Latexin Protein

A cDNA sequence encoding the Latexin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12541-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930759

  • 189.00€ / 20µg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Latexin Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The Latexin protein solution contains 20mM Tris-HCl, pH 7.5, 50mM NaCl and 10% Glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MEIPPTNYPA SRAALVAQNY INYQQGTPHR VFEVQKVKQA SMEDIPGRGH KYRLKFAVEE IIQKQVKVNC TAEVLYPSTG QETAPEVNFTFEGETGKNPD EEDNTFYQRL KSMKEPLEAQ NIPDNFGNVS PEMTLVLHLA WVACGYIIWQ NSTEDTWYKM VKIQTVKQVQ RNDDFIELDYTILLHNIASQ EIIPWQMQVL WHPQYGTKVK HNSRLPKEVQ LE
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions