missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LGALS16 Protein

A cDNA sequence encoding the LGALS16 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12561-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15940819

  • 212.00€ / 50µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LGALS16 Protein
Research Use Only
Recombinant Protein
E. coli
MSFLTVPYKL PVSLSVGSCV IIKGTLIDSS INEPQLQVDF YTEMNEDSEI AFHLRVHLGR RVVMNSREFG IWMLEENLHY VPFEDGKPFD LRIYVCLNEY EVKVNGEYIY AFVHRIPPSY VKMIQVWRDV SLDSVLVNNG RR
Greater than 95.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
1x PBS, pH 7.4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions