missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LGALS8 Protein

A cDNA sequence encoding the LGALS8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10768-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15925108

  • 189.00€ / 10µg

Estimated Shipment: 27-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

LGALS8 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MMLSLNNLQN IIYNPVIPFV GTIPDQLDPG TLIVIRGHVP SDADRFQVDL QNGSSMKPRA DVAFHFNPRF KRAGCIVCNT LINEKWGREE ITYDTPFKRE KSFEIVIMVL KDKFQVAVNG KHTLLYGHRI GPEKIDTLGI YGKVNIHSIG FSFSSDLQST QASSLELTEI SRENVPKSGT PQLRLPFAAR LNTPMGPGRT VVVKGEVNAN AKSFNVDLLA GKSKDIALHL NPRLNIKAFV RNSFLQESWG EEERNITSFP FSPGMYFEMI IYCDVREFKV AVNGVHSLEY KHRFKELSSI DTLEINGDIH LLEVRSW
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The ED50 of Galectin-8 as determined by its ability to agglutinate human red blood cells is 0.8∼4 microg/ml.
Human
Untagged
LGALS8 was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.4.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions