missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human LIF, Biotinylated Protein

Product Code. 18737417
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Unit Size:
25µg
Product Code. Formulation unitSize
18737417 Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein 25µg
18795688 Lyophilized from a 0.2μm filtered solution in PBS 25µg
2 options
This item is not returnable. View return policy

Product Code. 18737417

Brand: R&D Systems™ BT7734025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LIF, Biotinylated Protein is derived from E. coli. The Recombinant Human LIF, Biotinylated Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P15018
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Gene ID (Entrez) 3976
Molecular Weight (g/mol) 20 kDa
Quantity 25 μg
Source E. coli-derived human LIF protein Pro24-Phe202
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Biotinylated protein via amines
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Biological Activity 0.02 to 0.12ng/mL
Product Type Recombinant protein
Conjugate Biotin
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.