missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LILRB4/CD85k/ILT3 Fc Biotin Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human LILRB4/CD85k/ILT3 Fc Chimera binds to Human Fibronectin Protein (Catalog # 1918-FN ) with an ED50 of 0.400-4.00 μg/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 11006 |
| Molecular Weight (g/mol) | M.W.-theoretical: 53 kDa, M.W.-observed: 50-70 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human LILRB4/CD85k/ILT3 protein Human LILRB4 (Gly24-Glu259) (N-terminus) Accession # NP_001265355.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD85 antigen-like family member K, CD85k, CD85k antigen, HM18, ILT3, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5, LIR5LILRB5, LIR-5subfamily B, member 4, member 4 |
| Conjugate | Biotin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction