Learn More
R&D Systems™ Recombinant Human LRP-1 Cluster IV Fc Chimera Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Descripción
Measured by its binding ability in a functional ELISA. When Recombinant Human LRP-1 Cluster IV Fc Chimera (Catalog # 5395-L4B) is immobilized at 50 ng/mL (100 μL/well), Recombinant Human LRPAP (Catalog # 4296-LR ) binds with an ED50 of 0.0250-0.350 ng/mL.
Especificaciones
Especificaciones
| Accession Number | Q07954.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4035 |
| Molecular Weight (g/mol) | M.W. Observed: 100-120 kDa, under reducing conditions., M.W. Theoretical: 76.7 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human LRP-1 Cluster IV protein Human LRP1-C4 (Ser3332-Asp3779) (N-terminus) Accession # Q07954.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido