missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LTF Protein
Click to view available options
Quantity:
1 mg
10 μg
10 mg
2 μg
200 μg
50 μg
Unit Size:
10mg
10µg
1mg
200µg
2µg
50µg
Specifications
Specifications
| Gene ID (Entrez) | 4057 |
| Name | LTF Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Gene Symbol | LTF |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction