missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAK16 Protein

A cDNA sequence encoding the MAK16 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12630-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931049

  • 7875.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MAK16 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQSDDVI WDTLGNKQFC SFKIRTKTQS FCRNEYSLTG LCNRSSCPLA NSQYATIKEE KGQCYLYMKV IERAAFPRRL WERVRLSKNY EKALEQIDEN LIYWPRFIRH KCKQRFTKIT QYLIRIRKLT LKRQRKLVPL SKKVERREKR REEKALIAAQ LDNAIEKELL ERLKQDTYGD IYNFPIHAFD KALEQQEAES DSSDTEEKDD DDDDEEDVGK REFVEDGEVD ESDISDFEDM DKLDASSDED QDGKSSSEEE EEKALSAKHK GKMPLRGPLQ RKRAYVEIEY EQETEPVAKA KTT
Greater than 90.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MAK16 protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4), 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions