missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAOA Protein
A cDNA sequence encoding the MAOA was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | MAOA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931059
|
enQuireBio™
QP12633-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921059
|
enQuireBio™
QP12633-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911059
|
enQuireBio™
QP12633-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MAOA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAOA protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMENQEKA SIAGHMFDVV VIGGGISGLS AAKLLTEYGV SVLVLEARDR VGGRTYTIRN EHVDYVDVGG AYVGPTQNRI LRLSKELGIE TYKVNVSERL VQYVKGKTYP FRGAFPPVWN PIAYLDYNNL WRTIDNMGKE IPTDAPWEAQ HADKWDKMTM KELIDKICWT KTARRFAYLF VNINVTSEPH EVSALWFLWY VKQCGGTTRI FSVTNGGQER KFVGGSGQVS ERIMDLLGDQ VKLNHPVTHV DQSSDNIIIE TLNHEHYECK YVINAIPPTL TAKIHFRPEL PAERNQLIQR LPMGAVIKCM MYYKEAFWKK KDYCGCMIIE DEDAPISITL DDTKPDGSLP AIMGFILARK ADRLAKLHKE IRKKKICELY AKVLGSQEAL HPVHYEEKNW CEEQYSGGCY TAYFPPGIMT QYGRVIRQPV GRIFFAGTET ATKWSGYMEG AVEAGERAAR EVLNGLGKVT EKDIWVQEPE SKDVPAVEIT HTFWERNLPS | |
Greater than 80% as determined by SDS-PAGE. |