missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mast cell carboxypeptidase A Protein
A DNA sequence encoding the Homo sapiens (Human) Mast cell carboxypeptidase A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5856-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001861.2 | |
Mast cell carboxypeptidase A Protein | |
Research Use Only | |
CPA3 | |
Recombinant Protein | |
E. coli | |
IPGRHSYAKY NNWEKIVAWT EKMMDKYPEM VSRIKIGSTV EDNPLYVLKI GEKNERRKAI FTDCGIHARE WVSPAFCQWF VYQATKTYGR NKIMTKLLDR MNFYILPVFN VDGYIWSWTK NRMWRKNRSK NQNSKCIGTD LNRNFNASWN SIPNTNDPCA DNYRGSAPES EKETKAVTNF IRSHLNEIKV YITFHSYSQM LLFPYGYTSK LPPNHEDLAK VAKIGTDVLS TRYETRYIYG PIESTIYPIS GSSLDWAYDL GIKHTFAFEL RDKGKFGFLL PESRIKPTCR ETMLAVKFIA KYILKHTS | |
Greater than 90% as determined by SDS-PAGE. |
1359 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |