missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MAT1A Protein

Product Code. 15931109
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15931109

Brand: enQuireBio™ QP126491mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the MAT1A was constructed and used to recombinantly synthesize the protein.

Specifications

Name MAT1A Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHS SGLVPRGSHM NGPVDGLCDH SLSEGVFMFT SESVGEGHPD KICDQISDAV LDAHLKQDPN AKVACETVCK TGMVLLCGEI TSMAMVDYQR VVRDTIKHIG YDDSAKGFDF KTCNVLVALE QQSPDIAQCV HLDRNEEDVG AGDQGLMFGY ATDETEECMP LTIILAHKLN ARMADLRRSG LLPWLRPDSK TQVTVQYMQD NGAVIPVRIH TIVISVQHNE DITLEEMRRA LKEQVIRAVV PAKYLDEDTV YHLQPSGRFV IGGPQGDAGV TGRKIIVDTY GGWGAHGGGA FSGKDYTKVD RSAAYAARWV AKSLVKAGLC RRVLVQVSYA IGVAEPLSIS IFTYGTSQKT ERELLDVVHK NFDLRPGVIV RDLDLKKPIY QKTACYGHFG RSEFPWEVPR KLVF
Buffer The MAT1A protein solution contains 20mM Tris-HCl pH 8.0, 1mM DTT, 100mM NaCl and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.