missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAVS Protein
A cDNA sequence encoding the MAVS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12652-5ug
Additional Details : Weight : 0.01000kg
Specifications
MAVS Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPFAEDK TYKYICRNFS NFCNVDVVEI LPYLPCLTAR DQDRLRATCT LSGNRDTLWH LFNTLQRRPG WVEYFIAALR GCELVDLADE VASVYQSYQP RTSDRPPDPL EPPSLPAERP GPPTPAAAHS IPYNSCREKE PSYPMPVQET QAPESPGENS EQALQTLSPR AIPRNPDGGP LESSSDLAAL SPLTSSGHQE QDTELGSTHT AGATSSLTPS RGPVSPSVSF QPLARSTPRA SRLPGPTGSV VSTGTSFSSS SPGLASAGAA EGKQGAESDQ AEPIICSSGA EAPANSLPSK VPTTLMPVNT VALKVPANPA SVSTVPSKLP TSSKPPGAVP SNALTNPAPS KLPINSTRAG MVPSKVPTSM VLTKVSASTV PTDGSSRNEE TPAAPTPAGA TGGSSAWLDS SSENRGLGSE LSKPGVLASQ VDSPFSGCFE DLAISASTSL GMGPCHGPEE NEYKSEGTFG IHVAENPSIQ LLEGNPGPPA DPDGGPRPQA DRKFQEREVP CHRPSP | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAVS solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |