missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MEP1A Protein

A cDNA sequence encoding the MEP1A was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifications

Name MEP1A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15931199
View Documents Promotion Details
enQuireBio™
QP12678-1UG
1 μg
137.00€
1µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15951199
View Documents Promotion Details
enQuireBio™
QP12678-5UG
5 μg
212.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15941199
View Documents Promotion Details
enQuireBio™
QP12678-50UG
50 μg
1815.00€
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

MEP1A Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MEP1A protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Research Use Only
Recombinant Protein
Baculovirus
ADPVPIKYLP EENVHDADFG EQKDISEINL AAGLDLFQGD ILLQKSRNGL RDPNTRWTFP IPYILADNLG LNAKGAILYA FEMFRLKSCV DFKPYEGESS YIIFQQFDGC WSEVGDQHVG QNISIGQGCA YKAIIEHEIL HALGFYHEQS RTDRDDYVNI WWDQILSGYQ HNFDTYDDSL ITDLNTPYDY ESLMHYQPFS FNKNASVPTI TAKIPEFNSI IGQRLDFSAI DLERLNRMYN CTTTHTLLDH CTFEKANICG MIQGTRDDTD WAHQDSAQAG EVDHTLLGQC TGAGYFMQFS TSSGSAEEAA LLESRILYPK RKQQCLQFFY KMTGSPSDRL VVWVRRDDST GNVRKLVKVQ TFQGDDDHNW KIAHVVLKEE QKFRYLFQGT KGDPQNSTGG IYLDDITLTE TPCPTGVWTV RNFSQVLENT SKGDKLQSPR FYNSEGYGFG VTLYPNSRES SGYLRLAFHV CSGENDAILE WPVENRQVII TILDQEPDVR NRMSSSMVFT TSKSHTSPAI NDTVIWDRPS RVGTYHTDCN CFRSIDLGWS GFISHQMLKR RSFLKNDDLI IFVDFEDITH LSQHHHHHH
Greater than 85.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications