missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMP 8 Protein

Product Code. 15925178
Click to view available options
Quantity:
1 IU
10 IU
2.5 IU
Unit Size:
1 unit
10 units
2.5 units
This item is not returnable. View return policy

Product Code. 15925178

Brand: enQuireBio™ QP107891U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the MMP 8 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4317
Name MMP 8 Protein
Quantity 1 IU
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol MMP8
Biological Activity 100 units/ml after activation with APMA by solution assay method. One unit of collagenolytic activity is defined as the cleavage of 1microg of collagen per minute by the solution method.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MFSLKTLPFL LLLHVQISKA FPVSSKEKNT KTVQDYLEKF YQLPSNQYQS TRKNGTNVIV EKLKEMQRFF GLNVTGKPNE ETLDMMKKPR CGVPDSGGFM LTPGNPKWER TNLTYRIRNY TPQLSEAEVE RAIKDAFELW SVASPLIFTR ISQGEADINI AFYQRDHGDN SPFDGPNGIL AHAFQPGQGI GGDAHFDAEE TWTNTSANYN LFLVAAHEFG HSLGLAHSSD PGALMYPNYA FRETSNYSLP QDDIDGIQAI YGLSSNPIQP TGPSTPKPCD PSLTFDAITT LRGEILFFKD RYFWRRHPQL QRVEMNFISL FWPSLPTGIQ AAYEDFDRDL IFLFKGNQYW ALSGYDILQG YPKDISNYGF PSSVQAIDAA VFYRSKTYFF VNDQFWRYDN QRQFMEPGYP KSISGAFPGI ESKVDAVFQQ EHFFHVFSGP RYYAFDLIAQ RVTRVARGNK WLNCRYG
Buffer The MMP-8 protein solution (100 units/ml) in 0.05M Tris-HCl buffer, pH 7.6, 0.2M NaCl, 5mM CaCl2, 0.0025% NaN3 and 0.1% BSA.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.