missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP23B Protein
A cDNA sequence encoding the MMP23B was constructed and used to recombinantly synthesize the protein.
137.00€ - 5450.00€
Specifications
Name | MMP23B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941299
|
enQuireBio™
QP12709-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931299
|
enQuireBio™
QP12709-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921299
|
enQuireBio™
QP12709-1MG |
1 mg |
5450.00€
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MMP23B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MMP23B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSYTLTPAR LRWDHFNLTY RILSFPRNLL SPRETRRALA AAFRMWSDVS PFSFREVAPE QPSDLRIGFY PINHTDCLVS ALHHCFDGPT GELAHAFFPP HGGIHFDDSE YWVLGPTRYS WKKGVWLTDL VHVAAHEIGH ALGLMHSQHG RALMHLNATL RGWKALSQDE LWGLHRLYG | |
Greater than 85% as determined by SDS-PAGE. |