missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MMP28 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | MMP28 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSFAKQGNK WYKQHLSYRL VNWPEHLPEP AVRGAVRAAF QLWSNVSALE FWEAPATGPA DIRLTFFQGD HNDGLGNAFD GPGGALAHAF LPRRGEAHFD QDERWSLSRR RGRNLFVVLA HEIGHTLGLT HSPAPRALMA PYYKRLGRDA LLSWDDVLAV QSLYGKPLGG SVAVQLPGKL FTDFETWDSY SPQGRRPETQ GPKYCHSSFD AITVDRQQQL YIFKGSHFWE VAADGNVSEP RPLQERWVGL PPNIEAAAVS LNDGDFYFFK GGRCWRFRGP KPVWGLPQLC RAGGLPRHPD AALFFPPLRR LILFKGARYY VLARGGLQVE PYYPRSLQDW GGIPEEVSGA LPRPDGSIIF FRDDRYWRLD QAKLQATTSG RWATELPWMG CWHANSGSAL F |
| Buffer | The MMP28 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction