missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS2 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | MRPS2 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMATSSAA LPRILGAGAR APSRWLGFLG KATPRPARPS RRTLGSATAL MIRESEDSTD FNDKILNEPL KHSDFFNVKE LFSVRSLFDA RVHLGHKAGC RHRFMEPYIF GSRLDHDIID LEQTATHLQL ALNFTAHMAY RKGIILFISR NRQFSYLIEN MARDCGEYAH TRYFRGGMLT NARLLFGPTV RLPDLIIFLH TLNNIFEPHV AVRDAAKMNI PTVGIVDTNC NPCLITYPVP GNDDSPLAVH LYCRLFQTAI TRAKEKRQQV EALYRLQGQK EPGDQGPAHP PGADMSHSL |
| Buffer | The MRPS2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction