missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MVK Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | MVK Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMLSEVLL VSAPGKVILH GEHAVVHGKV ALAVSLNLRT FLRLQPHSNG KVDLSLPNIG IKRAWDVARL QSLDTSFLEQ GDVTTPTSEQ VEKLKEVAGL PDDCAVTERL AVLAFLYLYL SICRKQRALP SLDIVVWSEL PPGAGLGSSA AYSVCLAAAL LTVCEEIPNP LKDGDCVNRW TKEDLELINK WAFQGERMIH GNPSGVDNAV STWGGALRYH QGKISSLKRS PALQILLTNT KVPRNTRALV AGVRNRLLKF PEIVAPLLTS IDAISLECER VLGEMGEAPA PEQYLVLEEL IDMNQHHLNA LGVGHASLDQ LCQVTRARGL HSKLTGAGGG GCGITLLKPG LEQPEVEATK QALTSCGFDC LETSIGAPGV SIHSATSLDS RVQQALDGL |
| Buffer | MVK protein (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction