missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDE1 Protein
A cDNA sequence encoding the NDE1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | NDE1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931599
|
enQuireBio™
QP12805-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921599
|
enQuireBio™
QP12805-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911599
|
enQuireBio™
QP12805-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NDE1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NDE1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 5mM DTT and 0.2M NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEDSGKTFSS EEEEANYWKD LAMTYKQRAE NTQEELREFQ EGSREYEAEL ETQLQQIETR NRDLLSENNR LRMELETIKE KFEVQHSEGY RQISALEDDL AQTKAIKDQL QKYIRELEQA NDDLERAKRA TIMSLEDFEQ RLNQAIERNA FLESELDEKE NLLESVQRLK DEARDLRQEL AVQQKQEKPR TPMPSSVEAE RTDTAVQATG SVPSTPIAHR GPSSSLNTPG SFRRGLDDST GGTPLTPAAR ISALNIVGDL LRKVGALESK LASCRNLVYD QSPNRTGGPA SGRSSKNRDG GERRPSSTSV PLGDKGLDTS CRWLSKSTTR SSSSC | |
Greater than 85.0% as determined by SDS-PAGE. |