missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFA4 Protein

A cDNA sequence encoding the NDUFA4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12811-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15911629

  • 4090.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4697
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMLRQIIG QAKKHPSLIP LFVFIGTGAT GATLYLLRLA LFNPDVCWDR NNPEPWNKLG PNDQYKFYSV NVDYSKLKKE RPDF
Greater than 80% as determined by SDS-PAGE.
NDUFA4 Protein
Research Use Only
NDUFA4
Human
His
NDUFA4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions