missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB9 Protein
A cDNA sequence encoding the NDUFB9 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Gene ID (Entrez) | 4715 |
---|---|
Name | NDUFB9 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | NDUFB9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911649
|
enQuireBio™
QP12817-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991639
|
enQuireBio™
QP12817-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901649
|
enQuireBio™
QP12817-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4715 | |
Research Use Only | |
NDUFB9 | |
Human | |
His | |
The NDUFB9 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
NDUFB9 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAFLASG PYLTHQQKVL RLYKRALRHL ESWCVQRDKY RYFACLMRAR FEEHKNEKDM AKATQLLKEA EEEFWYRQHP QPYIFPDSPG GTSYERYDCY KVPEWCLDDW HPSEKAMYPD YFAKREQWKK LRRESWEREV KQLQEETPPG GPLTEALPPA RKEGDLPPLW WYIVTRPRER PM | |
Greater than 90.0% as determined by SDS-PAGE. |