missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Neutrophil defensin 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Neutrophil defensin 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5924-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035965.1 | |
Neutrophil defensin 1 Protein | |
Research Use Only | |
DEFA1 | |
Recombinant Protein | |
E. coli | |
DIPEVVVSLA WDESLAPKHP GSRKNMACYC RIPACIAGER RYGTCIYQGR LWAFCC | |
Greater than 80% as determined by SDS-PAGE. |
1667 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |