missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMT1 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | NMT1 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMADESET AVKPPAPPLP QMMEGNGNGH EHCSDCENEE DNSYNRGGLS PANDTGAKKK KKKQKKKKEK GSETDSAQDQ PVKMNSLPAE RIQEIQKAIE LFSVGQGPAK TMEEASKRSY QFWDTQPVPK LGEVVNTHGP VEPDKDNIRQ EPYTLPQGFT WDALDLGDRG VLKELYTLLN ENYVEDDDNM FRFDYSPEFL LWALRPPGWL PQWHCGVRVV SSRKLVGFIS AIPANIHIYD TEKKMVEINF LCVHKKLRSK RVAPVLIREI TRRVHLEGIF QAVYTAGVVL PKPVGTCRYW HRSLNPRKLI EVKFSHLSRN MTMQRTMKLY RLPETPKTAG LRPMETKDIP VVHQLLTRYL KQFHLTPVMS QEEVEHWFYP QENIIDTFVV ENANGEVTDF LSFYTLPSTI MNHPTHKSLK AAYSFYNVHT QTPLLDLMSD ALVLAKMKGF DVFNALDLME NKTFLEKLKF GIGDGNLQYY LYNWKCPSMG AEKVGLVLQ |
| Buffer | NMT1 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4). |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction