missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Noggin Fc Chimera Protein

Product Code. 18765937
Click to view available options
Quantity:
100 μg
50 μg
500 μg
Unit Size:
100µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18765937

Brand: R&D Systems™ 3344NG500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New and Improved Noggin is Now Available! The new protein has ∼2 fold better activity, higher purity, and no tag! Earn double rewards for reviews

Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.100-0.400 μg/mL in the presence of 30 ng/mL of Recombinant Human BMP-4 (Catalog number- 314-BP ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q13253
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 49.6 kDa (monomer)
Name Noggin Fc Chimera
Quantity 500 μg
Source Mouse myeloma cell line, NS0-derived human Noggin protein, (N-terminus) Human Noggin (Gln28-Cys232) Accession Number-Q13253 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Gene Symbol NOG
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.