missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG1-beta 1A Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_039250.2 |
| Name | NRG1-beta 1A Protein |
| Quantity | 50 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | NRG1 |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction