missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ORAOV1 Protein
A cDNA sequence encoding the ORAOV1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Name | ORAOV1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931989
|
enQuireBio™
QP12930-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911989
|
enQuireBio™
QP12930-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921989
|
enQuireBio™
QP12930-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ORAOV1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ORAOV1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAGSQD IFDAIVMADE RFHGEGYREG YEEGSSLGVM EGRQHGTLHG AKIGSEIGCY QGFAFAWKCL LHSCTTEKDS RKMKVLESLI GMIQKFPYDD PTYDKLHEDL DKIRGKFKQF CSLLNVQPDF KISAEGSGLS F | |
Greater than 90.0% as determined by SDS-PAGE. |