Learn More
DiscoverX Recombinant Human OX40L
Product Code. 17392619
Encompass
Click to view available options
:
10 μg
Product Code. 17392619
Brand: DiscoverX 921312
Recombinant Human OX40L is an agonist for OX40 (CD134). Provided as 10μg of dry powder alongside reconstitution buffer.
OX40 ligand (OX40L or gp34), a type II transmembrane glycoprotein that binds and acts as an agonist for CD134 (OX40, a member of the TNF receptor superfamily). It is one of the co-stimulatory molecules in the immune system and is speculated to promote the survival and proliferation of activated CD4+ T cells and prolong the immune response. Provided as 10μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 10 μg
Product Type Control Ligands