Learn More
enQuireBio™ Recombinant Human PBEF / Visfatin / NAMPT Protein
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
Specifications
Specifications
| Accession Number | NP_005737.1 |
| Gene ID (Entrez) | 10135 |
| Name | PBEF / Visfatin / NAMPT Protein |
| Quantity | 25 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | NAMPT |
| Biological Activity | Human PBEF / Visfatin / NAMPT induces IL-6 production in human PBMCs as detected via ELISA (typical ED50 is 100 ng/ml). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |