missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCBP1 Protein
A cDNA sequence encoding the PCBP1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | PCBP1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15922119
|
enQuireBio™
QP12972-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912119
|
enQuireBio™
QP12972-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15902119
|
enQuireBio™
QP12972-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PCBP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PCBP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDAGVTE SGLNVTLTIR LLMHGKEVGS IIGKKGESVK RIREESGARI NISEGNCPER IITLTGPTNA IFKAFAMIID KLEEDINSSM TNSTAASRPP VTLRLVVPAT QCGSLIGKGG CKIKEIREST GAQVQVAGDM LPNSTERAIT IAGVPQSVTE CVKQIC | |
Greater than 85.0% as determined by SDS-PAGE. |