missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF-AB Protein
Recombinant Protein
112.00€ - 5450.00€
Specifications
Name | PDGF-AB Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | PDGFA/B |
Biological Activity | Shows bioactivity at a concentration of 20 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935659
|
enQuireBio™
QP5201-2UG |
2 μg |
112.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915659
|
enQuireBio™
QP5201-10UG |
10 μg |
263.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905659
|
enQuireBio™
QP5201-100UG |
100 μg |
720.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945659
|
enQuireBio™
QP5201-500UG |
500 μg |
3330.00€
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925659
|
enQuireBio™
QP5201-1MG |
1 mg |
5450.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PDGF-AB Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
Research Use Only | |
PDGFA/B | |
Recombinant Protein | |
E. coli | |
Alphachain :MSIEEAVPA VCKTRTVIYE IPRSQVDPTS ANFLIWPPCV EVKRCTGCCN TSSVKCQPSR VHHRSVKVAK VEYVRKKPKL KEVQVRLEEH LECACATTSL NPDYREEDTG RPRESGKKRK RKRLKPT Bet achain:MSL GSLTIAEPAM IAECKTRTEV FEISRRLIDR TNANFLVWPP CVEVQRCSGC CNNRNVQCRP TQVQLRPVQV RKIEIVRKKP IFKKATVTLE DHLACKCETV AAARPVT | |
Greater than 95% as determined by SDS-PAGE. |