missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDPN Protein

A cDNA sequence encoding the PDPN was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name PDPN Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922199
View Documents Promotion Details
enQuireBio™
QP12999-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15912199
View Documents Promotion Details
enQuireBio™
QP12999-25UG
25 μg
212.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15902199
View Documents Promotion Details
enQuireBio™
QP12999-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PDPN Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein solution contains 20mM Tris-HCl pH 7.5 and 0.1M NaCl.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MASTGQPEDD TETTGLEGGV AMPGAEDDVV TPGTSEDRYK SGLTTLVATS VNSVTGIRIE DLPTSESTVH AQEQSPSATA SNVATSHSTE KVDGDTQTTV EKDGLSTVTL
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications