missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP3 Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase FKBP3, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_002004.1 |
---|---|
Gene ID (Entrez) | 2287 |
Name | Peptidyl-prolyl cis-trans isomerase FKBP3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979609
|
enQuireBio™
QP6049-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999609
|
enQuireBio™
QP6049-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989609
|
enQuireBio™
QP6049-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15909619
|
enQuireBio™
QP6049-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15919619
|
enQuireBio™
QP6049-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15929619
|
enQuireBio™
QP6049-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_002004.1 | |
Peptidyl-prolyl cis-trans isomerase FKBP3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2287 | |
Research Use Only | |
FKBP3 | |
Recombinant Protein | |
E. coli | |
AAAVPQRAWT VEQLRSEQLP KKDIIKFLQE HGSDSFLAEH KLLGNIKNVA KTANKDHLVT AYNHLFETKR FKGTESISKV SEQVKNVKLN EDKPKETKSE ETLDEGPPKY TKSVLKKGDK TNFPKKGDVV HCWYTGTLQD GTVFDTNIQT SAKKKKNAKP LSFKVGVGKV IRGWDEALLT MSKGEKARLE IEPEWAYGKK GQPDAKIPPN AKLTFEVELV DID | |
Greater than 90% as determined by SDS-PAGE. |