missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3K?D946GST Protein
A cDNA sequence encoding the PI3K?D946GST was constructed and used to recombinantly synthesize the protein.
212.00€ - 695.00€
Specifications
Name | PI3K?D946GST Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15912359
|
enQuireBio™
QP13051-1UG |
1 μg |
212.00€
1µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922359
|
enQuireBio™
QP13051-2.5UG |
2.5 μg |
389.00€
2.50µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932359
|
enQuireBio™
QP13051-5UG |
5 μg |
695.00€
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PI3K?D946GST Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
GST | |
Greater than 95.0% as determined by SDS Page. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
0.4 mg/ml solution in 25mM Tris-HCl, pH 8.0, 50mM NaCl, 0.5mM MgCl2, and 50% glycerol. |