missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PL 12 Protein

Product Code. 15942409
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15942409

Brand: enQuireBio™ QP130685ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PL 12 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PL 12 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence MDSTLTASEI RQRFIDFFKR NEHTYVHSSA TIPLDDPTLL FANAGMNQFK PIFLNTIDPS HPMAKLSRAA NTQKCIRAGG KHNDLDDVGK DVYHHTFFEM LGSWSFGDYF KELACKMALE LLTQEFGIPI ERLYVTYFGG DEAAGLEADL ECKQIWQNLG LDDTKILPGN MKDNFWEMGD TGPCGPCSEI HYDRIGGRDA AHLVNQDDPN VLEIWNLVFI QYNREADGIL KPLPKKSIDT GMGLERLVSV LQNKMSNYDT DLFVPYFEAI QKGTGARPYT GKVGAEDADG IDMAYRVLAD HARTITVALA DGGRPDNTGR GYVLRRILRR AVRYAHEKLN ASRGFFATLV DVVVQSLGDA FPELKKDPDM VKDIINEEEV QFLKTLSRGR RILDRKIQSL GDSKTIPGDT AWLLYDTYGF PVDLTGLIAE EKGLVVDMDG FEEERKLAQL KSQGKGAGGE DLIMLDIYAI EELRARGLEV TDDSPKYNYH LDSSGSYVFE NTVATVMALR REKMFVEEVS TGQECGVVLD KTCFYAEQGG QIYDEGYLVK VDDSSEDKTE FTVKNAQVRG GYVLHIGTIY GDLKVGDQVW LFIDEPRRRP IMSNHTATHI LNFALRSVLG EADQKGSLVA PDRLRFDFTA KGAMSTQQIK KAEEIANEMI EAAKAVYTQD CPLAAAKAIQ GLRAVFDETY PDPVRVVSIG VPVSELLDDP SGPAGSLTSV EFCGGTHLRN SSHAGAFVIV TEEAIAKGIR RIVAVTGAEA QKALRKAESL KKCLSVMEAK VKAQTAPNKD VQREIADLGE ALATAVIPQW QKDELRETLK SLKKVMDDLD RASKADVQKR VLEKTKQFID SNPNQPLVIL EMESGASAKA LNEALKLFKM HSPQTSAMLF TVDNEAGKIT CLCQVPQNAA NRGLKASEWV QQVSGLMDGK GGGKDVSAQA TGKNVGCLQE ALQLATSFAQ LRLGDVKN
Buffer PL-12 is supplied in16mM HEPES buffer pH 8.0, 250mM sodium chloride, and 20% glycerol.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.