missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PLA2G10 Protein

A cDNA sequence encoding the PLA2G10 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13071-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922419

  • 5400.00€ / 1mg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PLA2G10 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Sterile filtered and lyophilized from 0.5 mg/ml in 0.01M Tris buffer pH 7.2.
Greater than 95% as determined by SDS PAGE.
Research Use Only
Recombinant Protein
E. coli
MGPLPVCLPI MLLLLLPSLL LLLLLPGPGS GEASRILRVH RRGILELAGT VGCVGPRTPI AYMKYGCFCG LGGHGQPRDA IDWCCHGHDC CYTRAEEAGC SPKTERYSWQ CVNQSVLCGP AENKCQELLC KCDQEIANCL AQTEYNLKYL FYPQFLCEPD SPKCD
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions