missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POGLUT1 Protein

A cDNA sequence encoding the POGLUT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13090-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922479

  • 1665.00€ / 50µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

POGLUT1 Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The POGLUT1 solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
RQKESGSKWK VFIDQINRSL ENYEPCSSQN CSCYHGVIEE DLTPFRGGIS RKMMAEVVRR KLGTHYQITK NRLYRENDCM FPSRCSGVEH FILEVIGRLP DMEMVINVRD YPQVPKWMEP AIPVFSFSKT SEYHDIMYPA WTFWEGGPAV WPIYPTGLGR WDLFREDLVR SAAQWPWKKK NSTAYFRGSR TSPERDPLIL LSRKNPKLVD AEYTKNQAWK SMKDTLGKPA AKDVHLVDHC KYKYLFNFRG VAASFRFKHL FLCGSLVFHV GDEWLEFFYP QLKPWVHYIP VKTDLSNVQE LLQFVKANDD VAQEIAERGS QFIRNHLQMD DITCYWENLL SEYSKFLSYN VTRRKGYDQI IPKMLKTELL EHHHHHH
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions