missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPIF Protein
A cDNA sequence encoding the PPIF was constructed and used to recombinantly synthesize the protein.
137.00€ - 2725.00€
Specifications
Gene ID (Entrez) | 10105 |
---|---|
Name | PPIF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | PPIF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915298
|
enQuireBio™
QP10829-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935298
|
enQuireBio™
QP10829-50UG |
50 μg |
212.00€
50µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925298
|
enQuireBio™
QP10829-1MG |
1 mg |
2725.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10105 | |
Research Use Only | |
PPIF | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH CSKGSGDPSS SSSSGNPLVY LDVDANGKPL GRVVLELKAD VVPKTAENFR ALCTGEKGFG YKGSTFHRVI PSFMCQAGDF TNHNGTGGKS IYGSRFPDEN FTLKHVGPGV LSMANAGPNT NGSQFFICTI KTDWLDGKHV VFGHVKEGMD VVKKIESFGS KSGRTSKKIV ITDCGQLS | |
Greater than 95.0% as determined by SDS-PAGE. |
PPIF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 250 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
Human | |
His | |
PPIF solution containing 20mM Tris-HCl pH 7.5, 10% glycerol and 1mM DTT. |