missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPIL2 Protein

A cDNA sequence encoding the PPIL2 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Gene ID (Entrez) 23759
Name PPIL2 Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol PPIL2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15925308
View Documents Promotion Details
enQuireBio™
QP10832-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15905308
View Documents Promotion Details
enQuireBio™
QP10832-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915308
View Documents Promotion Details
enQuireBio™
QP10832-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

23759
Research Use Only
PPIL2
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGKRQHQKDK MYITCAEYTH FYGGKKPDLP QTNFRRLPFD HCSLSLQPFV YPVCTPDGIV FDLLNIVPWL KKYGTNPSNG EKLDGRSLIK LNFSKNSEGK YHCPVLFTVF TNNTHIVAVR TTGNVYAYEA VEQLNIKAKN FRDLLTDEPF SRQDIITLQD PTNLDKFNVS NFYHVKNNMK IIDPDEEKAK QDPSYYLKNT NAETRETLQE YKEFKGDEI LAATMKAPEK KKVDKLNAAH YSTGKVSASF TSTAMVPETT EAAAIDEDV LRYQFVKKKG YVRLHTNKGD LNLELHCDLT PKTCENFIRL CKKHYYDGTI FHRSIRNFVI QGGDPTGTGT GGESYWGKPF KDEFRPNLSH TGRGILSMAN SGPNSNRSQF FITFRSCAYL DKKHTIFGRV VGGFDVLTAM ENVESDPKTD RPKEEIRIDA TTVFVDPYEE ADAQIAQERK TQLKVAPETK VKSSQPQAGS QGPQTFRQGV GKYINPAATE QQRKSPQPVP LSPCPRRSPV GVLGTSAPGS SRLPDDH
Greater than 95% as determined by SDS-PAGE.
PPIL2 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 290 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin.
Human
His
The PPIL2 solution contains 20mM Tris-HCl pH 8.0, 0.1M NaCl and 20% glycerol.
Videos
SDS
Documents

Documents

Product Certifications