missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRDX3 Protein

A cDNA sequence encoding the PRDX3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10839-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935328

  • 125.00€ / 5µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PRDX3 Protein
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MPAVTQHAPY FKGTAVVNGE FKDLSLDDFK GKYLVLFFYP LDFTFVCPTE IVAFSDKANE FHDVNCEVVA VSVDSHFSHL AWINTPRKNG GLGHMNIALL SDLTKQISRD YGVLLEGSGL ALRGLFIIDP NGVIKHLSVN DLPVGRSVEE TLRLVKAFQY VETHGEVCPA NWTPDSPTIK PSPAASKEYF QKVNQ
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The specific activity was found to be approximately 82-83 pmole/min/microg. The enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX3 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for 1 minute.
Human
Untagged
The PRDX3 solution contains 20mM Tris-HCl pH 8.0, and 10% glycerol.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions