missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRKRA Protein

A cDNA sequence encoding the PRKRA was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name PRKRA Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922649
View Documents Promotion Details
enQuireBio™
QP13146-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15902649
View Documents Promotion Details
enQuireBio™
QP13146-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15912649
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP13146-1MG
1 mg
7875.00€
1mg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PRKRA Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PRKRA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSQSRHR AEAPPLERED SGTFSLGKMI TAKPGKTPIQ VLHEYGMKTK NIPVYECERS DVQIHVPTFT FRVTVGDITC TGEGTSKKLA KHRAAEAAIN ILKANASICF AVPDPLMPDP SKQPKNQLNP IGSLQELAIH HGWRLPEYTL SQEGGPAHKR EYTTICRLES FMETGKGASK KQAKRNAAEK FLAKFSNISP ENHISLTNVV GHSLGCTWHS LRNSPGEKIN LLKRSLLSIP NTDYIQLLSE IAKEQGFNIT YLDIDELSAN GQYQCLAELS TSPITVCHGS GISCGNAQSD AAHNALQYLK IIAERK
Greater than 80.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates