missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKRA Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | PRKRA Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSQSRHR AEAPPLERED SGTFSLGKMI TAKPGKTPIQ VLHEYGMKTK NIPVYECERS DVQIHVPTFT FRVTVGDITC TGEGTSKKLA KHRAAEAAIN ILKANASICF AVPDPLMPDP SKQPKNQLNP IGSLQELAIH HGWRLPEYTL SQEGGPAHKR EYTTICRLES FMETGKGASK KQAKRNAAEK FLAKFSNISP ENHISLTNVV GHSLGCTWHS LRNSPGEKIN LLKRSLLSIP NTDYIQLLSE IAKEQGFNIT YLDIDELSAN GQYQCLAELS TSPITVCHGS GISCGNAQSD AAHNALQYLK IIAERK |
| Buffer | PRKRA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol, 2mM DTT and 1mM EDTA. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction