missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Dr1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Dr1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5946-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001929.1 | |
Protein Dr1 Protein | |
Research Use Only | |
DR1 | |
Recombinant Protein | |
E. coli | |
ASSSGNDDDL TIPRAAINKM IKETLPNVRV ANDARELVVN CCTEFIHLIS SEANEICNKS EKKTISPEHV IQALESLGFG SYISEVKEVL QECKTVALKR RKASSRLENL GIPEEELLRQ QQELFAKARQ QQAELAQQEW LQMQQAAQQA QLAAASASAS NQAGSSQDEE DDDDI | |
Greater than 80% as determined by SDS-PAGE. |
1810 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |