missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRPS2 Protein

A cDNA sequence encoding the PRPS2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13170-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932719

  • 189.00€ / 20µg

Estimated Shipment: 27-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PRPS2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
PRPS2 protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPNIVLF SGSSHQDLSQ RVADRLGLEL GKVVTKKFSN QETSVEIGES VRGEDVYIIQ SGCGEINDNL MELLIMINAC KIASSSRVTA VIPCFPYARQ DKKDKVGESR APISAKLVAN MLSVAGADHI ITMDLHASQI QGFFDIPVDN LYAEPAVLQW IRENIAEWKN CIIVSPDAGG AKRVTSIADR LNVEFALIHK ERKKANEVDR MVLVGDVKDR VAILVDDMAD TCGTICHAAD KLLSAGATKV YAILTHGIFS GPAISRINNA AFEAVVVTNT IPQEDKMKHC TKIQVIDISM ILAEAIRRTH NGESVSYLFS HVPL
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions