missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PSMD9 Protein

A cDNA sequence encoding the PSMD9 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name PSMD9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922799
View Documents Promotion Details
enQuireBio™
QP13194-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15912799
View Documents Promotion Details
enQuireBio™
QP13194-20UG
20 μg
212.00€
20µg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15902799
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP13194-1MG
1 mg
4090.00€
1mg
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PSMD9 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The PSMD9 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSDEEAR QSGGSSQAGV VTVSDVQELM RRKEEIEAQI KANYDVLESQ KGIGMNEPLV DCEGYPRSDV DLYQVRTARH NIICLQNDHK AVMKQVEEAL HQLHARDKEK QARDMAEAHK EAMSRKLGQS ESQGPPRAFA KVNSISPGSP ASIAGLQVDD EIVEFGSVNT QNFQSLHNIG SVVQHSEGKP LNVTVIRRGE KHQLRLVPTR WAGKGLLGCN IIPLQR
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates