missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTHrP Protein

A cDNA sequence encoding the PTHrP was constructed and used to recombinantly synthesize the protein.

137.00€ - 2055.00€

Specifications

Name PTHrP Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15922829
View Documents Promotion Details
enQuireBio™
QP13204-10UG
10 μg
137.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15942829
View Documents Promotion Details
enQuireBio™
QP13204-50UG
50 μg
212.00€
50µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15932829
View Documents Promotion Details
enQuireBio™
QP13204-1MG
1 mg
2055.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PTHrP Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
PthRp protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4.
Research Use Only
Recombinant Protein
E. coli
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications