missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RARRES1 Protein

A cDNA sequence encoding the RARRES1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13270-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933039

  • 189.00€ / 25µg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

RARRES1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RARRES1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSPDDPGQP QDAGVPRRLL QQAARAALHF FNFRSGSPSA LRVLAEVQEG RAWINPKEGC KVHVVFSTER YNPESLLQEG EGRLGKCSAR VFFKNQKPRP TINVTCTRLI EKKKRQQEDY LLYKQMKQLK NPLEIVSIPD NHGHIDPSLR LIWDLAFLGS SYVMWEMTTQ VSHYYLAQLT SVRQWKTNDD TIDFDYTVLL HELSTQEIIP CRIHLVWYPG KPLKVKYHCQ ELQTPEEASG TEEGSAVVPT ELSNF
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions