missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBBP4 Protein
A cDNA sequence encoding the RBBP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13276-20ug
Additional Details : Weight : 0.01000kg
Specifications
RBBP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMADKEA AFDDAVEERV INEEYKIWKK NTPFLYDLVM THALEWPSLT AQWLPDVTRP EGKDFSIHRL VLGTHTSDEQ NHLVIASVQL PNDDAQFDAS HYDSEKGEFG GFGSVSGKIE IEIKINHEGE VNRARYMPQN PCIIATKTPS SDVLVFDYTK HPSKPDPSGE CNPDLRLRGH QKEGYGLSWN PNLSGHLLSA SDDHTICLWD ISAVPKEGKV VDAKTIFTGH TAVVEDVSWH LLHESLFGSV ADDQKLMIWD TRSNNTSKPS HSVDAHTAEV NCLSFNPYSE FILATGSADK TVALWDLRNL KLKLHSFESH KDEIFQVQWS PHNETILASS GTDRRLNVWD LSKIGEEQSP EDAEDGPPEL LFIHGGHTAK ISDFSWNPNE PWVICSVSED NIMQVWQMAE NIYNDEDPEG SVDPEGQGS | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RBBP4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM EDTA. |