missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBBP4 Protein

Product Code. 15923059
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15923059

Brand: enQuireBio™ QP132765ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the RBBP4 was constructed and used to recombinantly synthesize the protein.

Specifications

Name RBBP4 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSHMADKEA AFDDAVEERV INEEYKIWKK NTPFLYDLVM THALEWPSLT AQWLPDVTRP EGKDFSIHRL VLGTHTSDEQ NHLVIASVQL PNDDAQFDAS HYDSEKGEFG GFGSVSGKIE IEIKINHEGE VNRARYMPQN PCIIATKTPS SDVLVFDYTK HPSKPDPSGE CNPDLRLRGH QKEGYGLSWN PNLSGHLLSA SDDHTICLWD ISAVPKEGKV VDAKTIFTGH TAVVEDVSWH LLHESLFGSV ADDQKLMIWD TRSNNTSKPS HSVDAHTAEV NCLSFNPYSE FILATGSADK TVALWDLRNL KLKLHSFESH KDEIFQVQWS PHNETILASS GTDRRLNVWD LSKIGEEQSP EDAEDGPPEL LFIHGGHTAK ISDFSWNPNE PWVICSVSED NIMQVWQMAE NIYNDEDPEG SVDPEGQGS
Buffer RBBP4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM EDTA.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.