missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RCAN1 Protein

A cDNA sequence encoding the RCAN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13286-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933089

  • 189.00€ / 20µg

Estimated Shipment: 27-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

RCAN1 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 7.5, 100mM NaCl and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKLYFAQTLH IGSSHLAPPN PDKQFLISPP ASPPVGWKQV EDATPVINYD LLYAISKLGPGEKYELHAAT DTTPSVVVHV CESDQEKEEE EEMERMRRPK PKIIQTRRPE YTPIHLS
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions