missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RCN2 Protein
A cDNA sequence encoding the RCN2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13289-1mg
Additional Details : Weight : 0.01000kg
Specifications
RCN2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEELHYPLGE RRSDYDREAL LGVQEDVDEY VKLGHEEQQK RLQAIIKKID LDSDGFLTES ELSSWIQMSF KHYAMQEAKQ QFVEYDKNSD DTVTWDEYNI QMYDRVIDFD ENTALDDAEE ESFRKLHLKD KKRFEKANQD SGPGLSLEEF IAFEHPEEVDYMTEFVIQEA LEEHDKNGDG FVSLEEFLGD YRWDPTANED PEWILVEKDR FVNDYDKDND GRLDPQELLP WVVPNNQGIA QEEALHLIDE MDLNGDKKLS EEEILENPDL FLTSEATDYG RQLHDDYFYH DEL | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RCN2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |